Learn More
Fisherbrand™ Drosophila Products, BuzzPlugs™ Vial and Bottle Closures
Nonshedding, dense weave of nontoxic cellulose acetate minimizes evaporation, keeps food fresh longer
Brand: Fisherbrand™ AS275
1083.97 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Nonshedding, dense weave of nontoxic cellulose acetate minimizes evaporation, keeps food fresh longer
- Superior barrier to mite infestation
- Exact fit with narrow 0.9 in. (25mm) or wide 1.1 in. (28.5mm) dia. plastic shell vials or standard (≈24mm dia.) glass vials or plastic stock bottles
- Stretches yet maintains tight fit after repeated removal and closing
- Flat surface accepts marking pen
Specifications
Minimizes evaporation, keeps food fresh longer | |
Superior barrier | |
Cellulose Acetate |
2.8cm wide plastic shell vials | |
1000 Case | |
BuzzPlugs Bottle Closure |