Learn More
Hirschmann™ Automatic Burettes, Acc. to Dr. Schilling
Click to view available options
Capacity (Metric):
5 mL
10 mL
15 mL
25 mL
50 mL
Unit Size:
Each
Description
- With Schellbach stripe
- Blue graduation
- Fast flow burette
Specifications
Specifications
Capacity (Metric) | 10 mL |
Material | Glass |
Graduations | 0.05 mL |
Includes | LDPE Bottle |
Scale | Blue |
Tolerance | ±0.05 mL |
Type | Schilling Buret |
Certifications/Compliance | DKD Certificate |