Learn More
Biochrom Disposable Visible/UV Cuvette
For use with UV-Visible spectrophotometers.
Brand:  Biochrom 80-3000-77
1315.75 NOK valid until 2025-08-24
Use promo code "24117" to get your promotional price.
 View more versions of this product
Product Code. 10166714
  • 1385.00 NOK / Pack of 100
Genomics Powerhouse
Use promo code 24117 at checkout
View Available Promotions
Only null left
Estimated Shipment: 17-10-2025
Description
  • Disposable cell for visible and UV spectrophotometry
  • Pathlength: 10mm
Specifications
Specifications
UV Visible Cuvet
800 μL
100 Pack
GeneQuant 1300 visible/UV spectrophotometer
Single-use
UV Plastic
10 mm