Learn More
Kartell™ Disposable Petri Dishes
Designed for routine lab work, bacteriological research
Brand: Kartell™ 0036700
1691.75 NOK valid until 2025-12-31
Use promo code "24345" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
CE marked, IVD | |
Clear | |
Polystyrene | |
Routine lab work, bacteriological research | |
65 mm |
Aspetic | |
Single-use | |
Contact Dish | |
1000 |