Learn More
Lovibond™ Comparator Disc
Lovibond™ Comparator Disc uses an array of glass fade-free color match standards for chemical test color comparison.
Brand: Lovibond™ 283680
2227.45 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Lovibond™ Comparator Disc uses an array of glass fade-free color match standards for chemical test color comparison.
- Easy to use, visual assessment of the sample; simply rotate disc to match color
- Glass, fade-free standards for longevity of use
- Shock resistant, scratch resistant
- Optional lighting units for guaranteed lighting conditions 24 hours/day
- Optional Nessleriser systems for longer path lengths
- More than 400 color discs are available for the Comparator 2000+
- Lovibond™ color standards consist exclusively of through-and-through colored and flat-ground glasses
Specifications
Dissolved Oxygen | |
Comparator 2000+ | |
Color Comparator Disc |
Disc NOE | |
0, 0.005, 0.01, 0.015, 0.03, 0.055, 0.08, 0.1, 0.12 mL/L |