Learn More
Grant Instruments™ Dimpled Mat for Platform Shaker
For use with PMR-30 Compact Fixed Angle Platform Rocker. Grant Instruments™ Dimpled Mat keeps vessels from sliding during operation.
Brand:  Grant Instruments™ PDM
Product Code. 11441038
  • 564.00 NOK / Each
Only null left
Estimated Shipment: 06-08-2025
Description
  • For use with PMR-30 Compact Fixed Angle Platform Rocker.
  • Keeps vessels from sliding during operation.
  • Dimensions: 215 x 215mm
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PMR-30