missing translation for 'onlineSavingsMsg'
Learn More

Diethyl(3-pyridyl)borane, 97%, Thermo Scientific Chemicals

Product Code. 15575266
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89878-14-8
Molecular Formula C9H14BN
Molecular Weight (g/mol) 147.03
MDL Number MFCD00012348
InChI Key OJKBCQOJVMAHDX-UHFFFAOYSA-N
Synonym diethyl 3-pyridyl borane, 3-diethylboryl pyridine, diethyl 3-pyridinyl borane, pyridine, 3-diethylboryl, 3-diethylboranyl pyridine, diethyl-3-pyridylborane, dithyl 3-pyridyl borane, 3-pyridyldiethylborane, diethyl 3-pyridyl bo
PubChem CID 642851
IUPAC Name diethyl(pyridin-3-yl)borane
SMILES CCB(CC)C1=CN=CC=C1

Specifications

Melting Point 149.0°C to 151.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 147.03
Percent Purity 97%
Chemical Name or Material Diethyl(3-pyridyl)borane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.