Learn More
Dicyclohexylphenylphosphine, 98%, Thermo Scientific Chemicals
CAS: 6476-37-5 | C18H27P | 274.39 g/mol
945.00 NOK - 3135.00 NOK
Chemical Identifiers
CAS | 6476-37-5 |
---|---|
Molecular Formula | C18H27P |
Molecular Weight (g/mol) | 274.39 |
MDL Number | MFCD00003854 |
InChI Key | VPLLTGLLUHLIHA-UHFFFAOYSA-N |
Synonym | dicyclohexylphenylphosphine, dicyclohexyl phenyl phosphine, phosphine, dicyclohexylphenyl, phenyldicyclohexylphosphine, phenylphosphinodicyclohexane, dicyclohexyl phenyl phosphane, pphcy2, acmc-1bey7, dicyclohexyl-phenyl-phosphane |
PubChem CID | 80970 |
IUPAC Name | dicyclohexyl(phenyl)phosphane |
SMILES | C1CCC(CC1)P(C1CCCCC1)C1=CC=CC=C1 |
Description
Dicyclohexylphenylphosphine, is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDicyclohexylphenylphosphine, is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Air Sensitive.
Chemical Identifiers
6476-37-5 | |
274.39 | |
VPLLTGLLUHLIHA-UHFFFAOYSA-N | |
80970 | |
C1CCC(CC1)P(C1CCCCC1)C1=CC=CC=C1 |
C18H27P | |
MFCD00003854 | |
dicyclohexylphenylphosphine, dicyclohexyl phenyl phosphine, phosphine, dicyclohexylphenyl, phenyldicyclohexylphosphine, phenylphosphinodicyclohexane, dicyclohexyl phenyl phosphane, pphcy2, acmc-1bey7, dicyclohexyl-phenyl-phosphane | |
dicyclohexyl(phenyl)phosphane |
Specifications
6476-37-5 | |
C18H27P | |
Air Sensitive | |
Insoluble in water. | |
C1CCC(CC1)P(C1CCCCC1)C1=CC=CC=C1 | |
274.39 | |
274.38 | |
Dicyclohexylphenylphosphine |
58°C to 61°C | |
MFCD00003854 | |
dicyclohexylphenylphosphine, dicyclohexyl phenyl phosphine, phosphine, dicyclohexylphenyl, phenyldicyclohexylphosphine, phenylphosphinodicyclohexane, dicyclohexyl phenyl phosphane, pphcy2, acmc-1bey7, dicyclohexyl-phenyl-phosphane | |
VPLLTGLLUHLIHA-UHFFFAOYSA-N | |
dicyclohexyl(phenyl)phosphane | |
80970 | |
98% |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 229-334-7
missing translation for 'rtecsNumber' : SY9125000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures