Learn More
Dichloromethylphosphine, 90%, Tech., Thermo Scientific Chemicals
Chemical Identifiers
CAS | 676-83-5 |
---|---|
Molecular Formula | CH3Cl2P |
Molecular Weight (g/mol) | 116.909 |
MDL Number | MFCD00013616 |
InChI Key | CDPKWOKGVUHZFR-UHFFFAOYSA-N |
Synonym | methyldichlorophosphine, dichloromethylphosphine, methylphosphonous dichloride, dichloro methyl phosphane, phosphonous dichloride, p-methyl, ch3pcl2, phosphonous dichloride, methyl, methyl phosphonous dichloride, methylphosphorus dichloride, phosphine, dichloromethyl |
PubChem CID | 61194 |
IUPAC Name | dichloro(methyl)phosphane |
SMILES | CP(Cl)Cl |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
676-83-5 | |
116.909 | |
CDPKWOKGVUHZFR-UHFFFAOYSA-N | |
61194 | |
CP(Cl)Cl |
CH3Cl2P | |
MFCD00013616 | |
methyldichlorophosphine, dichloromethylphosphine, methylphosphonous dichloride, dichloro methyl phosphane, phosphonous dichloride, p-methyl, ch3pcl2, phosphonous dichloride, methyl, methyl phosphonous dichloride, methylphosphorus dichloride, phosphine, dichloromethyl | |
dichloro(methyl)phosphane |
Specifications
676-83-5 | |
90% | |
CH3PCl2 | |
methyldichlorophosphine, dichloromethylphosphine, methylphosphonous dichloride, dichloro methyl phosphane, phosphonous dichloride, p-methyl, ch3pcl2, phosphonous dichloride, methyl, methyl phosphonous dichloride, methylphosphorus dichloride, phosphine, dichloromethyl | |
CP(Cl)Cl | |
116.909 | |
116.92 | |
Dichloromethylphosphine |
1.3000g/mL | |
CH3Cl2P | |
MFCD00013616 | |
CDPKWOKGVUHZFR-UHFFFAOYSA-N | |
dichloro(methyl)phosphane | |
61194 | |
Technical |
Safety and Handling
GHS H Statement
Harmful if swallowed. Harmful if inhaled. Causes severe skin burns and eye damage. Catches fire spontaneously if exposed to air. Reacts violently with water. Contact with water liberates toxic gas.
GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Do not allow contact with air.
Danger
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.