Learn More
Macherey-Nagel™ DiaQuant™ Peracetic Acid
Quick and easy check of residual paracetic acid in rinse water.
Brand: Macherey-Nagel™ 932001
467.95 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Specific to peracetic acid and it will not detect peroxides.
- The easy Dip and Read procedure provides reliable results in 10 seconds.
- Technicians may clean hemodialysis equipment using peracetic acid containing reagents (for example, Puristeril plus).
- Prior to the next use, they must rinse the system, until the reagent is completely washed out.
- Provides a quick, easy and dependable method to check rinse water and ensure safe dialysis.
Specifications
White to dark blue color change | |
100 Test Strips/Pk. | |
Colorimetric, semiquantitative test strip |
100 Test Strips/Pk. | |
0, 5, 10, 20, 30, 50 mg/L PAA |