Learn More
Epredia™ Cytospin™ 4 Cytofunnel™ Starter Kit
Accessory Starter Kit for the Epredia™ Cytospin™ 4 Cytocentrifuge. (Centrifuge is sold separately, Cat. No. A78300003)
8120.00 NOK - 19045.00 NOK
Specifications
For Use With (Equipment) Cytospin™ 4 Cytocentrifuge
View More Specs
Products 2
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
12097139
View Documents Promotion Details
Epredia™
A78710013
EZ Cytofunnel Starter Kit
8120.00 NOK
Each
View alternative products
Save Up to 
»
12017159
View Documents Promotion Details
Epredia™
3120110
Clipped Funnel Starter Kit
19045.00 NOK
Each
View alternative products
Save Up to 
»
Description
Description
The Cytofunnel Starter Kit provides everything needed for new Cytospin 4 users.
Available in both standard clipped Cytofunnel and EZ Cytofunnel options:
  • Clipped Cytofunnel Starter Kit
    • Includes: 50 Single White Cytofunnels, 25 Double Cytofunnels, Cytoslides, 12 Cytoclips
  • EZ Cytofunnel Starter Kit
    • Includes: 25 EZ Megafunnels, 20 EZ Single White Cytofunnels, 20 EZ Double Cytofunnels and Cytoslides
Specifications
Specifications
Cytospin™ 4 Cytocentrifuge