missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CytokeRatin 18 Mouse anti-Human, CoraLite 555, Clone: 1G11C4, Proteintech
Mouse Monoclonal Antibody
1696.32 NOK - 5264.45 NOK
Specifications
Antigen | CytokeRatin 18 |
---|---|
Clone | 1G11C4 |
Concentration | 1000 μg/mL |
Applications | Flow Cytometry, Immunofluorescence |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17761359
|
Proteintech
CL555-66187-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17751359
|
Proteintech
CL555-66187-100UL |
100 μL |
5264.45 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CytokeRatin 18 Monoclonal antibody specifically detects CytokeRatin 18 in Human samples. It is validated for FC (Intra), IFSpecifications
CytokeRatin 18 | |
1000 μg/mL | |
Monoclonal | |
Liquid | |
Human | |
3875 | |
Primary | |
Store at -20°C |
1G11C4 | |
Flow Cytometry, Immunofluorescence | |
CoraLite 555 | |
Mouse | |
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
IgG1 | |
Protein A purification | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title