Learn More
Cytochrome c Mouse anti-Human, CoraLite 594, Clone: 2D8D11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-66264-100UL
Description
The Cytochrome c antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Cytochrome c. This antibody recognizes human, mouse, rat antigen. The Cytochrome c antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™594.
Specifications
Cytochrome c | |
Monoclonal | |
2000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
2D8D11 | |
CoraLite 594 | |
100 μL | |
54205 | |
IgG2a |