Learn More
CYP24A1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21582-1-AP-150UL
 View more versions of this product
Product Code. 16861185
  • 4796.50 NOK / 150µL
Description
The CYP24A1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human CYP24A1. This antibody recognizes human, mouse, rat antigen. The CYP24A1 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
Specifications
CYP24A1
Polyclonal
Unconjugated
150 μL
1591
IgG
Western Blot, Immunoprecipitation, Immunohistochemistry
400 μg/mL
Rabbit
Primary
Human, Rat, Mouse