Learn More
Custom Select siRNA, In Vivo Ready
Custom Select siRNA, In Vivo Ready
153200.00 NOK
Specifications
Content And Storage | Contains dried siRNA and Nuclease-free Water (1.75 mL), shipped at ambient temperature. Upon receipt, store siRNAs in a non-frost-free freezer at or below –20 °C |
---|---|
RNAi Type | siRNA |
Product Type | siRNA |
Description
- Silencer™ Select custom siRNAs (LNA-modified 21-bp duplex with overhangs) are RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types.
- These siRNAs provide high knock-down efficiencies and specificities.
Specifications
Contains dried siRNA and Nuclease-free Water (1.75 mL), shipped at ambient temperature. Upon receipt, store siRNAs in a non-frost-free freezer at or below –20 °C | |
siRNA | |
siRNA |