Learn More
Sampling Systems™ Cup Liquid Sampler
Constructed of stainless steel and for sampling liquids and suspensions. Sampling Systems™ Cup Liquid Sampler enables discrete samples to be easily taken from a height within the product.
3990.00 - 8370.00
Product Code | Brand | Capacity (Metric) | Length (Metric) | Diameter (Metric) Outer | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Length (Metric) | Diameter (Metric) Outer | Price | Quantity & Availability | |||
11930018
|
Sampling Systems™
5030A-1000 |
100 mL | 1000 mm | 55 mm |
4165.00
Each |
|
|||
11920018
|
Sampling Systems™
5030A-600 |
100 mL | 600 mm | 55 mm |
3990.00
Each |
|
|||
11935437
|
Sampling Systems™
5030J-1000 |
500 mL | 1000 mm | 80 mm |
8160.00
Each |
|
|||
11945437
|
Sampling Systems™
5030J-1500 |
500 mL | 1500 mm | 80 mm |
8370.00
Each |
|
|||
Description
- 316L Stainless Steel Construction
- Single point liquid sampler
Specifications
Liquid Sampler | |
Stainless Steel |