Learn More
CPLX3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | CPLX3 |
---|---|
Concentration | 0.2 mg/mL |
Applications | Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16856744
|
Proteintech
16949-1-AP-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16846744
|
Proteintech
16949-1-AP-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Positively regulates a late step in synaptic vesicle exocytosis.Specifications
CPLX3 | |
Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q8R1B5, Q8WVH0 | |
235415, 501005, 594855 | |
CPLX3 Fusion Protein Ag10571 | |
Primary | |
-20°C | |
CPLX3 |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
complexin 3, Complexin III, CPLX3, CPX III, CPXIII, Nbla11589 | |
CPLX3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |