Learn More
Corning™ Reusable Plastic Erlenmeyer Flasks with Red Graduations
Translucent polymethylpentene; with polypropylene screw cap
Brand: Corning™ 4990P-50
This item is not returnable.
View return policy
Description
Translucent polymethylpentene flasks are manufactured with red graduations for reference. The flasks have a screw neck and are supplied with a PP cap.
Specifications
Polypropylene | |
Polypropylene | |
Narrow Mouth | |
1/Pk. |
50 mL | |
10 mL | |
6/Cs. | |
Screw Cap |