Learn More
Corning™ Reusable Plastic Erlenmeyer Flasks with Red Graduations
Translucent polymethylpentene; with polypropylene screw cap
Brand:  Corning™ 4990P-1L
 View more versions of this product
Product Code. 17203554
  • 1115.25 SEK / Pack of 4
Only null left
Estimated Shipment: 11-11-2025
Description
Translucent polymethylpentene flasks are manufactured with red graduations for reference. The flasks have a screw neck and are supplied with a PP cap.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 L
200 mL
4/Cs.
Polypropylene
Narrow Mouth
1/Pk.