Learn More
Corning™ 96-Well Next Generation CrystalEX Protein Crystallization Microplates
Optimized for high-throughput sitting-drop protein crystallization
7035.00 NOK - 9071.12 NOK
Specifications
Barcode | Alphanumeric |
---|---|
Lid | Without Lid |
Array | 8 x 12 |
Color | Clear |
No. of Wells | 96 |
Product Code | Brand | Well Shape | Volume (Metric) Well | Material | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Well Shape | Volume (Metric) Well | Material | Price | Quantity & Availability | |||
10471963
![]() |
Corning™
3550 |
Conical Flat | 1 μL | Polymer |
9071.12 NOK
Pack of 50 |
Estimated Shipment: 11-09-2025
Log in to see stock. |
|||
10057412
|
Corning™
3552 |
Round, Flat Bottom (F-well) | 2 μL | PZero | N/A | ||||
10436593
|
Corning™
3553 |
Conical Flat | 2 μL | PZero |
9071.12 NOK
Pack of 50 |
Estimated Shipment: 06-11-2025
Log in to see stock. |
|||
10327152
![]() |
Corning™
3556 |
Round | 4 μL | Cycloolefin Copolymer | N/A | ||||
Description
- Corning™ 96-well and 384-well crystallization microplates are optimized for high throughput protein crystal growth and screening
- Designed for sitting drop applications
- Meets ANSI/SBS standard footprint dimensions for microplates
- Ideal for fully automated crystal screening
- Multiple formats and versatility for custom options to maximize crystal formation, identification and analysis, and harvesting - Choose from four unique protein well shapes
- Cyclic olefin copolymer (COC) offers strong chemical compatibility and good optical clarity
- Reservoir numbers are embossed on each individual well for easy identification
- Options include one or three protein wells per reservoir well
Specifications
Alphanumeric | |
8 x 12 | |
96 | |
Non-treated | |
10/Pk. |
Without Lid | |
Clear | |
Non-sterile | |
50/Cs. |