Learn More
Julabo™ CORIO CD-Series Heating Circulators with Open, Stainless Steel Baths and Pump Connections
CORIO CD-Series Heating Circulators with Open, Stainless Steel Baths and Pump Connections are designed primarily for internal temperature control directly within the bath, however, external closed systems may be connected via the integrated pump connections.
18930.00 NOK - 39505.00 NOK
Specifications
Description | Circulation Capacity Flow Rate: 15 L/min., Circulation Capacity Pressure: 0.35 bar |
---|---|
Flow Rate | 15 L/min. |
Temperature Range | 20°C to 150°C (Working) |
Type | Heating Circulator with Open Bath |
For Use With (Application) | Internal and external applications at working temperatures up to +150°C |
Product Code | Brand | Model | Capacity (Metric) | Dimensions (L x W x H) | Dimensions (LxWxH) Interior | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Capacity (Metric) | Dimensions (L x W x H) | Dimensions (LxWxH) Interior | Price | Quantity & Availability | ||
15450572
![]() |
Julabo™
9012405 |
CD-B5 | 3.5 to 5 L | 38 x 23 x 41 cm | 15 x 15 x 15 cm (Usable Bath Opening) |
18930.00 NOK
Each |
Estimated Shipment: 23-09-2025
Log in to see stock. |
||
15470572
![]() |
Julabo™
9012417 |
CD-B17 | 13 to 17 L | 40 x 38 x 47 cm | 18 x 30 x 20 cm (Usable Bath Opening) |
22825.00 NOK
Each |
Estimated Shipment: 23-09-2025
Log in to see stock. |
||
15430582
![]() |
Julabo™
9012439 |
CD-B39 | 35 to 41 L | 34 x 54 x 57 cm | 30 x 33 x 30 cm (Usable Bath Opening) |
39505.00 NOK
Each |
Estimated Shipment: 17-09-2025
Log in to see stock. |
||
Description
- Bright, White, Easy to Read Display
- Easy to Operate Front Controls
- Easy Switching Between Internal and External Circulation
- Low Liquid Level Early Warning System
- Integrated Drain Screw (except CD-B5)
- Integrated Pump Connection (1 x M16)
- Integrated Timer
- PID1 and ACC Controls
- Very Quiet
Specifications
Circulation Capacity Flow Rate: 15 L/min., Circulation Capacity Pressure: 0.35 bar | |
20°C to 150°C (Working) | |
Internal and external applications at working temperatures up to +150°C | |
0.35 bar |
15 L/min. | |
Heating Circulator with Open Bath | |
2 kW | |
±0.03°C |