missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated RBAP48 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66060-20UL
This item is not returnable.
View return policy
Description
The RBAP48 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RBAP48. This antibody recognizes human antigen. The RBAP48 antibody has been validated for the following applications: IF/ICC, FC (Intra) analysis. This antibody is coralite™ plus 488.Specifications
RBAP48 | |
Monoclonal | |
2000 μg/mL | |
NURF55; RBAP48 | |
Recombinant Protein | |
20 μL | |
Primary | |
Human | |
53 kDa | |
Liquid |
Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
5C4D6 | |
CoraLite Plus 488 | |
Mouse | |
Protein A purification | |
RUO | |
5928 | |
Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
Conjugated Antibody | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction