missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated DRP1 (C-terminal) Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-12957-20UL
This item is not returnable.
View return policy
Description
The DRP1 (C-terminal) antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human DRP1 (C-terminal). This antibody recognizes human, mouse, rat antigen. The DRP1 (C-terminal) antibody has been validated for the following applications: IF/ICC, FC (Intra) analysis. This antibody is coralite™ plus 488.Specifications
DRP1 (C-terminal) | |
Polyclonal | |
CoraLite Plus 488 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
10059 | |
Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
Conjugated Antibody | |
IgG |
Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
1000 μg/mL | |
DLP1; DRP1; DVLP; DYMPLE; DYNIV-11; FLJ41912; HDYNIV; VPS1 | |
Recombinant Protein | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
75-80 kDa | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction