missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated C1QA Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67063-20UL
This item is not returnable.
View return policy
Description
The C1QA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human C1QA. This antibody recognizes human antigen. The C1QA antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.Specifications
C1QA | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Protein G purification | |
RUO | |
712 | |
Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
Conjugated Antibody | |
IgG1 |
Immunocytochemistry/Immunofluorescence | |
1B10E3 | |
CoraLite Plus 488 | |
Recombinant Protein | |
20 μL | |
Primary | |
Human | |
30 kDa | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction