Learn More
Contec™ VertiKlean™ Sox Mop Head Covers
Increases durability of VertiKlean mop when used on abrasive surfaces. Contec™ VertiKlean™ Sox Mop Head Covers are available in regular, medium and large to fit all VertiKlean mop heads.
1635.00 NOK - 8600.00 NOK
Specifications
Product Type | Mop Cover |
---|---|
Sterility | Non-sterile |
Autoclavable | Not Autoclavable |
Disposable | Single-use |
For Use With (Application) | Removable cover increases durability of VertiKlean mop when used on abrasive surfaces |
Product Code | Brand | Size | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | For Use With (Equipment) | Price | Quantity & Availability | ||||
11976935
![]() |
Contec™
VK-SOX |
Regular | Regular VertiKlean Mop Head | ||||||
11966935
![]() ![]() |
Contec™
MVK-QT-SOX |
Medium | Medium VertiKlean Mop Head |
8600.00 NOK
Pack of 120 |
Estimated Shipment: 16-10-2025
Log in to see stock. |
||||
11956935
![]() ![]() |
Contec™
LVK-SOX |
Large | Large VertiKlean Mop Head |
2505.00 NOK
Pack of 32 |
Estimated Shipment: 16-10-2025
Log in to see stock. |
||||
Description
- Clean-processed polyester
- Extends the life of VertiKlean mop heads
Specifications
Mop Cover | |
Not Autoclavable | |
Removable cover increases durability of VertiKlean mop when used on abrasive surfaces |
Non-sterile | |
Single-use | |
Polyester |