Learn More
Contec™ VertiKlean™ MAX™ Mop Heads
Clean walls, ceilings and other hard-to-reach surfaces with ease. VertiKlean™ MAX™ Mop Heads easily snap onto frames and last longer than traditional mops.
816.90 NOK - 7905.00 NOK
Specifications
Product Type | Mop Head |
---|---|
Autoclavable | Not Autoclavable |
Disposable | Single-use |
Product Code | Brand | Dimensions | Sterility | Material | Head Type | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions | Sterility | Material | Head Type | Price | Quantity & Availability | ||
13408166
|
Contec™
VKMX0200 |
32 x 11 x 2 cm | Non-sterile | Polyester, Foam | Non-sealed Edge | ||||
15184589
|
Contec™
VKMX0250 |
32 x 11 x 2 cm | Sterile | Polyester, Foam | Non-sealed Edge | ||||
16605564
![]() ![]() |
Contec™
VKMX0125 |
21 x 11 x 2 cm | Non-sterile | Ribbed Polyester Microfiber, Foam | Non-sealed Edge |
5380.00 NOK
Pack of 48 |
Estimated Shipment: 05-12-2025
Log in to see stock. |
||
16615564
![]() ![]() |
Contec™
VKMX0175 |
21 x 11 x 2 cm | Sterile | Ribbed Polyester Microfiber, Foam | Non-sealed Edge |
7905.00 NOK
Pack of 48 |
Estimated Shipment: 05-12-2025
Log in to see stock. |
||
15684937
![]() ![]() |
Contec™
VKMX0225 |
32 x 11 x 2 cm | Non-sterile | Ribbed Polyester Microfiber, Foam | Non-sealed Edge |
3750.00 NOK
Pack of 28 |
Estimated Shipment: 20-11-2025
Log in to see stock. |
||
15585040
![]() ![]() |
Contec™
VKMX0275 |
32 x 11 x 2 cm | Sterile | Ribbed Polyester Microfiber, Foam | Non-sealed Edge |
5545.00 NOK
Pack of 28 |
Estimated Shipment: 10-09-2025
Log in to see stock. |
||
15610496
![]() ![]() |
Contec™
VKMX0325 |
38 x 11 x 2 cm | Non-sterile | Ribbed Polyester Microfiber, Foam | Non-sealed Edge |
5240.00 NOK
Each |
Estimated Shipment: 05-12-2025
Log in to see stock. |
||
16625564
![]() ![]() |
Contec™
VKMX0375 |
38 x 11 x 2 cm | Sterile | Ribbed Polyester Microfiber, Foam | Non-sealed Edge |
6175.00 NOK
Pack of 24 |
Estimated Shipment: 05-12-2025
Log in to see stock. |
||
15646996
![]() ![]() |
Contec™
VKSE0200 |
38 x 13 x 1 cm | Non-sterile | Ribbed Polyester Microfiber, Foam | Ribbed Sealed Edge |
3650.00 NOK
Pack of 16 |
Estimated Shipment: 29-08-2025
Log in to see stock. |
||
15656996
![]() ![]() |
Contec™
VKSE0250 |
38 x 13 x 1 cm | Sterile | Ribbed Polyester Microfiber, Foam | Ribbed Sealed Edge |
6205.00 NOK
Pack of 24 |
Estimated Shipment: 26-09-2025
Log in to see stock. |
||
16348845
![]() ![]() |
Contec™
VKSE0100 |
24 x 13 x 1 cm | Non-sterile | Ribbed Polyester Microfiber, Hydrophilic Urethane Foam | Sealed Edge |
5755.00 NOK
Pack of 40 |
Estimated Shipment: 05-09-2025
Log in to see stock. |
||
16358845
![]() ![]() |
Contec™
VKSE0150 |
24 x 13 x 1 cm | Sterile | Ribbed Polyester Microfiber, Hydrophilic Urethane Foam | Sealed Edge |
7230.00 NOK
Pack of 40 |
Estimated Shipment: 29-10-2025
Log in to see stock. |
||
16426830
![]() ![]() |
Contec™
VKSE0300 |
46 x 13 x 1 cm | Non-sterile | Ribbed Polyester Microfiber, Hydrophilic Urethane Foam | Sealed Edge |
5085.00 NOK
Pack of 20 |
Estimated Shipment: 08-12-2025
Log in to see stock. |
||
16635564
![]() ![]() |
Contec™
VKSE0350 |
46 x 13 x 1 cm | Sterile | Ribbed Polyester Microfiber, Hydrophilic Urethane Foam | Sealed Edge |
7645.00 NOK
Pack of 20 |
Estimated Shipment: 05-12-2025
Log in to see stock. |
||
Description
- Features a single layer of hydrophilic polyurethane foam laminated to textured polyester microfiber fabric
- Backed by a semi-rigid thermoformed plate that easily snaps onto mop head frame
- Designed for applying disinfectants to both horizontal and vertical surfaces
- Mop heads are composed of a disposable absorbent sponge laminated to filament polyester knit fabric and backed with a thermoformed plate that easily snaps onto VertiKlean mop hardware
- Now available with ribbed microfiber for better contamination removal
- Pivoting mop head frame is ideal for cleaning walls, floors, and ceilings as well as hard-to-reach areas such as fume hoods, mini environments, and around light fixtures and heavy equipment
- Sterile mop heads are validated sterile per AAMI ISO11137 guidelines
- Autoclaving mop heads is not recommended due to thermoformed plate; however, all hardware is autoclavable
Order Info
Frames, handles and bucket wringer system sold separately.
Certifications
Validated sterile to a Sterility Assurance Level of 10-6 per AAMI ISO11137:2006 Guidelines. Certificate of sterility is available.
Specifications
Mop Head | |
Not Autoclavable | |
Single-use |