Learn More
Gilson™ Connection Kit
2630.00 NOK - 2920.00 NOK
Specifications
Quantity 5 Set
Type Connection Kit
View More Specs
Products 2
Product Code Brand Diameter (Metric) Outside Price Quantity & Availability  
Product Code Brand Diameter (Metric) Outside Price Quantity & Availability  
10404332
View Documents
Gilson™
F1825056
4 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10113002
View Documents
Gilson™
F1825054
3 mm
2920.00 NOK
Pack of 5
Only null left
Estimated Shipment: 10-09-2025
Add to basket
Description
Description
  • Connector kit accessories for the MINIPULS™ 3 and Evolution peristaltic pumps come in a variety of types: metal sleeve connectors for small bore tubing, connector kits for flanged tubing, or plastic connectors for large bore tubing.
  • These kits contain five connectors each.
  • The connectors are available in a three sizes to fit a variety of tubing of different inner diameters.
Specifications
Specifications
5 Set
Connection Kit