Learn More
Greiner Bio-One Conical Bottom Polystyrene Test Tubes with Bayonet Cap
For centrifugation and applications that require chemical and thermo resistant vessels.
€ 458.67 - € 596.27
Specifications
Capacity (Metric) | 12 mL |
---|---|
Length (Metric) | 100 mm |
Material | Polystyrene |
For Use With | Centrifugation and applications that require chemical and thermo resistant vessels. |
Includes | Skirted bottom |
Product Code | Brand | Sterility | Closure Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sterility | Closure Color | Price | Quantity & Availability | ||||
10707492
|
Greiner Bio-One
164180 |
Non-sterile | White |
€ 596.27
Pack of 1300 |
Estimated Shipment: 06-08-2025
Log in to see stock. |
||||
10324561
![]() |
Greiner Bio-One
164161 |
Sterile | Blue | ||||||
Description
- Volume: 12mL
- Skirted bottom
- Dia. X Height: 16.8 x 100mm
Specifications
12 mL | |
Polystyrene | |
Skirted bottom | |
Bayonet Cap |
100 mm | |
Centrifugation and applications that require chemical and thermo resistant vessels. | |
16.8 mm | |
Test Tube |