Learn More
Buerkle™ Compact Jerry Can and Compact Stopcock Set
Set features tamper-proof, polypropylene closure material.
708.00 NOK - 824.00 NOK
Specifications
Closure Material | Polypropylene |
---|---|
Closure Type | Tamper-proof |
Color | Transparent |
Description | Space saving jerry can with spigot |
Graduations | Moulded litre |
Product Code | Brand | Capacity (Metric) | Dimensions (L x W x H) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Dimensions (L x W x H) | Price | Quantity & Availability | ||||
11971451
|
Buerkle™
0435-3000 |
5 L | 335 x 65 x 335 mm |
708.00 NOK
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
||||
15833954
|
Buerkle™
0435-3010 |
10 L | 335 x 125 x 335 mm |
824.00 NOK
Each |
Estimated Shipment: 11-07-2025
Log in to see stock. |
||||
Description
- PP, transparent
- Tamper-proof closure
- Autoclavable canister (without valve)
- Molded liter graduation
- Aluminum canister support accssory
Specifications
Polypropylene | |
Transparent | |
Moulded litre | |
Spigot Compact | |
Polypropylene | |
Non-sterile |
Tamper-proof | |
Space saving jerry can with spigot | |
335 mm | |
335 mm | |
S50 Thread |