Learn More
OHAUS™ Column Mounts
OHAUS™ Column Mounts are designed for use with a range of Defender™ 5000 Bases.
1350.00 NOK - 2695.00 NOK
Specifications
Languages | Chinese, English (US), French, German, Italian, Japanese, Korean, Polish, Portuguese (BR), Russian, Spanish (MX) |
---|---|
Product Line | Column Kit |
Product Code | Brand | Type | Weight (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Weight (Metric) | Price | Quantity & Availability | ||||
15938465
|
OHAUS™
30424410 |
CS D52xx | 2.300 kg |
N/A
|
N/A | ||||
15948465
|
OHAUS™
30424411 |
CS D52xx | 4.100 kg |
1350.00 NOK
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15958465
|
OHAUS™
30424412 |
CS D52xx | 6.200 kg |
1525.00 NOK
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15968465
|
OHAUS™
30424413 |
SST D52xx | 2.300 kg |
1445.00 NOK
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15978465
|
OHAUS™
30424414 |
SST D52xx | 4.100 kg |
2375.00 NOK
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15988465
|
OHAUS™
30424415 |
SST D52xx | 6.200 kg |
2695.00 NOK
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For use with a base
Specifications
Chinese, English (US), French, German, Italian, Japanese, Korean, Polish, Portuguese (BR), Russian, Spanish (MX) | |
Column Kit |