Learn More
Ansell MICROFLEX™ Cobalt™ N19 Nitrile Gloves
Multi-purpose nitrile gloves offer consistent comfort and durability to workers across a wide range of industries. Fully textured with a 4 mil thickness helps wearers securely grip tools and materials.
22865.00 NOK
Specifications
AQL | 1.5 |
---|---|
Color | Blue |
Disposable | Single-use |
Finger Thickness (English) | 4.3 mil |
Finger Thickness (Metric) | 0.11 mm |
Description
- Soft and stretchy nitrile material
- Durable and puncture resistant
- Fully textured surface for enhanced gripping in wet or dry conditions
Specifications
1.5 | |
Single-use | |
0.11 mm | |
Nitrile | |
0.10 mm | |
4 mil | |
20.5 MPa | |
24.13 cm |
Blue | |
4.3 mil | |
Yes | |
3.9 mil | |
Powder Free | |
0.10 mm | |
9.5 in. |