Learn More
Fisherbrand™ Clear Graduated Glass Bottles with White Polypropylene Caps
Fisherbrand™ Clear Graduated Glass Bottles with White Polypropylene Caps feature a wide mouth for easy removal of contents.
324.00 NOK - 2245.00 NOK
Specifications
Color | Clear |
---|---|
Material | Glass (Type 3) |
Closure Color | White |
Closure Included | Yes |
Graduated | Yes |
Product Code | Brand | Capacity (Metric) | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Quantity | Price | Quantity & Availability | ||||
16124377
![]() |
Fisherbrand™
FB02911453A |
30 mL | 48 |
1385.00 NOK
Pack of 48 |
Estimated Shipment: 28-08-2025
Log in to see stock. |
||||
16134377
![]() |
Fisherbrand™
FB02911454A |
60 mL | 48 |
539.00 NOK
Pack of 48 |
Estimated Shipment: 02-01-2026
Log in to see stock. |
||||
16164387
![]() |
Fisherbrand™
FB02911467A |
120 mL | 144 |
1885.00 NOK
Pack of 144 |
Estimated Shipment: 22-09-2025
Log in to see stock. |
||||
16144377
![]() |
Fisherbrand™
FB02911455A |
120 mL | 24 |
324.00 NOK
Pack of 24 |
Estimated Shipment: 01-10-2025
Log in to see stock. |
||||
16174387
![]() |
Fisherbrand™
FB02911468A |
240 mL | 96 |
2245.00 NOK
Pack of 96 |
Estimated Shipment: 13-02-2026
Log in to see stock. |
||||
16154377
![]() |
Fisherbrand™
FB02911456A |
240 mL | 24 |
673.00 NOK
Pack of 24 |
Estimated Shipment: 02-10-2025
Log in to see stock. |
||||
16184387
![]() |
Fisherbrand™
FB02911469A |
480 mL | 48 |
1310.00 NOK
Pack of 48 |
Estimated Shipment: 01-12-2025
Log in to see stock. |
||||
16164377
![]() |
Fisherbrand™
FB02911457A |
480 mL | 24 |
718.00 NOK
Pack of 24 |
Estimated Shipment: 24-09-2025
Log in to see stock. |
||||
Description
- Graduated in both milliliters and ounces
- Clear glass bottles are designed with a slight shoulder for easy pouring and handling
- Wide mouth facilitates easy mixing, handling, and transfer of contents
Specifications
Clear | |
White | |
Yes | |
Graduated Bottle | |
Polypropylene |
Glass (Type 3) | |
Yes | |
Wide | |
Pulp/Vinyl |