Learn More
Fisherbrand™ Clear Glass Alpha Bottles
Constructed of type 3 glass with a pilfer-proof screwed top. Fisherbrand™ Clear Glass Alpha Bottles are used mainly in pharmacological and laboratory procedures.
415.00 NOK - 462.00 NOK
Specifications
Color | Clear |
---|---|
Diameter (Metric) Inner Neck | 28 mm |
Material | Glass (Type 3) |
Product Code | Brand | Capacity (Metric) | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Quantity | Price | Quantity & Availability | ||||
15363408
|
Fisherbrand™
300779 |
30 mL | 156Pack |
462.00 NOK
Pack of 156 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||
15373408
|
Fisherbrand™
300773 |
60 mL | 127Pack |
415.00 NOK
Pack of 127 |
Estimated Shipment: 22-07-2025
Log in to see stock. |
||||
Description
- Lightweight
- Clear Glass
- Screwed top
Specifications
Clear | |
28 mm | |
Glass (Type 3) |