Learn More
Zwiebel™ Class M1 Weight Set
Use with weighing instruments of medium accuracy class III. Zwiebel™ Class M1 Weights are ideal for the verification or calibration of class M2 weights.
163.00€ - 1175.00€
Specifications
For Use With (Application) | For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII |
---|
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
11830962
|
Zwiebel™
PBI56845 |
1 to 500 g |
610.00€
Each |
Estimated Shipment: 26-08-2025
Log in to see stock. |
|||||
11850952
|
Zwiebel™
PBI5683W |
1 to 200 g |
425.00€
Each |
Estimated Shipment: 19-09-2025
Log in to see stock. |
|||||
11810982
|
Zwiebel™
PBI5684Y |
1 to 2000 g |
1175.00€
Each |
Estimated Shipment: 12-09-2025
Log in to see stock. |
|||||
11880922
|
Zwiebel™
PBI5682T |
1 to 500 mg |
163.00€
Each |
Estimated Shipment: 19-09-2025
Log in to see stock. |
|||||
11860932
|
Zwiebel™
PBI56835 |
1 to 50 g |
180.00€
Each |
Estimated Shipment: 19-09-2025
Log in to see stock. |
|||||
Description
- Class M2 stainless steel calibration weight
- Delivered with a calibration certificate guaranteeing traceability
- In compliance with OIML R111
- Supplied individually or as a set, in a plastic case or a wooden box
Specifications
For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII |