Learn More
Zwiebel™ Class F2 Box Weights
256.00 NOK - 1605.00 NOK
Specifications
For Use With (Application) | For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II |
---|
Product Code | Brand | Mass | Tolerance | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Tolerance | Price | Quantity & Availability | ||||
11840912
|
Zwiebel™
PBI56829 |
10 g | ±0.6 mg |
328.00 NOK
Each |
Estimated Shipment: 29-09-2025
Log in to see stock. |
||||
11870912
|
Zwiebel™
PBI5682C |
100 g | ±1.6 mg |
407.00 NOK
Each |
Estimated Shipment: 10-09-2025
Log in to see stock. |
||||
11800922
|
Zwiebel™
PBI5682F |
1000 g | ±15 mg |
1165.00 NOK
Each |
Estimated Shipment: 02-10-2025
Log in to see stock. |
||||
11810912
|
Zwiebel™
PBI56825 |
1 g | ±0.3 mg |
256.00 NOK
Each |
Estimated Shipment: 02-10-2025
Log in to see stock. |
||||
11810922
|
Zwiebel™
PBI5682H |
2000 g | ±30 mg |
1605.00 NOK
Each |
Estimated Shipment: 25-09-2025
Log in to see stock. |
||||
11880912
|
Zwiebel™
PBI5682D |
200 g | ±3 mg |
516.00 NOK
Each |
Estimated Shipment: 25-09-2025
Log in to see stock. |
||||
11820912
|
Zwiebel™
PBI56827 |
2 g | ±0.4 mg |
267.00 NOK
Each |
Estimated Shipment: 02-10-2025
Log in to see stock. |
||||
11890912
|
Zwiebel™
PBI5682E |
500 g | ±7.5 mg |
912.00 NOK
Each |
Estimated Shipment: 02-10-2025
Log in to see stock. |
||||
Specifications
For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II |