Learn More
Zwiebel™ Class E2 Box Weight
283.00 NOK - 1110.00 NOK
Specifications
For Use With (Application) | For use with Weighing Instruments of Special Accuracy Class I |
---|
Product Code | Brand | Mass | Tolerance | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Tolerance | Price | Quantity & Availability | ||||
11780623
|
Zwiebel™
PBI2302B |
10 mg | ±0.008 mg |
283.00 NOK
Each |
Estimated Shipment: 02-10-2025
Log in to see stock. |
||||
11720673
|
Zwiebel™
PBI23063 |
10 g | ±0.06 mg |
648.00 NOK
Each |
Estimated Shipment: 02-10-2025
Log in to see stock. |
||||
11730673
|
Zwiebel™
PBI23064 |
20 g | ±0.08 mg |
733.00 NOK
Each |
Estimated Shipment: 06-10-2025
Log in to see stock. |
||||
11760673
|
Zwiebel™
PBI23067 |
200 g | ±0.3 mg |
1110.00 NOK
Each |
Estimated Shipment: 23-09-2025
Log in to see stock. |
||||
11700673
|
Zwiebel™
PBI23061 |
2 g | ±0.04 mg |
576.00 NOK
Each |
Estimated Shipment: 02-10-2025
Log in to see stock. |
||||
11790663
|
Zwiebel™
PBI23060 |
1 g | ±0.03 mg |
543.00 NOK
Each |
Estimated Shipment: 02-10-2025
Log in to see stock. |
||||
11750623
|
Zwiebel™
PBI2301Z |
2 mg | ±0.006 mg |
283.00 NOK
Each |
Estimated Shipment: 02-10-2025
Log in to see stock. |
||||
11730633
|
Zwiebel™
PBI2302H |
500 mg | ±0.025 mg |
283.00 NOK
Each |
Estimated Shipment: 02-10-2025
Log in to see stock. |
||||
11710673
|
Zwiebel™
PBI23062 |
5 g | ±0.05 mg |
615.00 NOK
Each |
Estimated Shipment: 02-10-2025
Log in to see stock. |
||||
11740673
|
Zwiebel™
PBI23065 |
50 g | ±0.1 mg |
831.00 NOK
Each |
Estimated Shipment: 02-10-2025
Log in to see stock. |
||||
Specifications
For use with Weighing Instruments of Special Accuracy Class I |