Learn More
Afora™ Class A C/T Cylinders
71.00€ - 94.00€
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
12053433
|
Afora™
A6049 |
100 mL |
71.00€
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
|||||
12003433
|
Afora™
A6046/AM |
10 mL |
94.00€
Each |
Estimated Shipment: 14-08-2025
Log in to see stock. |
|||||
12083433
|
Afora™
A6050 |
250 mL |
92.00€
Each |
Estimated Shipment: 14-08-2025
Log in to see stock. |
|||||
Specifications
Glass | |
A | |
Cylinder |