Learn More
BRAND™ Class A Amber Borosilicate Volumetric Flask with Glass Stopper
Amber volumetric flask with glass stopper and USP certificate.
Brand:  BRAND™ 237464
931.46 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 16357997
  • 959.00 NOK / Pack of 2
Only null left
Estimated Shipment: 15-07-2025
Description
  • Borosilicate 3.3
  • To DIN EN ISO 1042
  • Calibrated to contain (TC, In)
  • Includes USP certificate, single, single batch, or batch delivery
TRUSTED_SUSTAINABILITY
Specifications
Specifications
To Contain
25 mL
Autoclavable
Yes
Non-sterile
USP Single Batch
Borosilicate Glass (3.3)
Reusable
Narrow
Flat