Learn More
Honeywell™ Retaining Covers for Prefilter Pads
Click to view available options
Cartridge Type:
Bayonet Gas/Vapor Cartridges on N-Series Respirators
Click-Fit Cartridges on T-Series Respirators
Pre-Assembled Cartridges on Freedom Series Respirators
Design Type:
Requires Honeywell™ Prefilter Pads [11847171 (P1) / 12682743 (P2)]
Requires Honeywell™ Prefilter Pads [7506P2 (Particulate, P2)]
For Use With (Equipment):
Honeywell™ Freedom Half-Mask Air-Purifying Respirators
Honeywell™ Half-Mask (e.g. N5500™, N7700™) and Full-Face (e.g. N5400™) Air-Purifying Respirators
Honeywell™ Half-Mask (e.g. Valuair™, Premier™, and MX/PF™) and Full-Face (e.g. Optifit Twin™) Air-Purifying Respirators
Unit Size:
1 pair
Each
Pack of 2
Pack of 20
Description
- Product Type: Retaining Cover
- Color: White
- Material: Plastic
- Requires: Prefilter Pad
Specifications
Specifications
Cartridge Type | Bayonet Gas/Vapor Cartridges on N-Series Respirators |
Color | White |
Design Type | Requires Honeywell™ Prefilter Pads [7506P2 (Particulate, P2)] |
Product Type | Retainer Cover for Prefilter Pads |
For Use With (Equipment) | Honeywell™ Half-Mask (e.g. N5500™, N7700™) and Full-Face (e.g. N5400™) Air-Purifying Respirators |
Includes | Spider and Retainer |
Model | N750036A |
Weight (Metric) | 907.6 g |