Learn More
OHAUS™ Clamp Connector
OHAUS™ Clamp Connector provides secure support for holding glassware at elevated positions.
Brand: OHAUS™ 30392262
451.55 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Hold glassware safely
- Corrosion-resistant
- Providing convenience with exceptional stability and durability
Specifications
Multi-Rod | |
Nickel-plated | |
40 x 33 mm (H x L) |
Connector | |
Clamp, Connector, Multi-Rod, CLC-MTRODZ |