Learn More
Macherey-Nagel™ Chromabond™ HR-XCW Polypropylene SPE Column
928.00 NOK - 3090.00 NOK
Specifications
Base Material | Spherical PS/DVB Copolymer |
---|---|
Column Type | SPE |
Particle Size | 85 μm |
Sorbent | Chromabond HR-XCW |
Type | Solid Phase Extraction Column |
Product Code | Brand | Column Capacity (Metric) | Pore Size | Description | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Column Capacity (Metric) | Pore Size | Description | Price | Quantity & Availability | |||
12777720
|
Macherey-Nagel™
730733 |
1 mL | 50 to 60 μm | Adsorbent Weight: 100mg |
1215.00 NOK
Pack of 30 |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||
12767720
|
Macherey-Nagel™
730731 |
1 mL | 50 to 60 μm | Adsorbent Weight: 30mg |
928.00 NOK
Pack of 30 |
In Stock
Log in to see stock. |
|||
12708212
|
Macherey-Nagel™
730739 |
3 mL | 50 to 60 μm | Adsorbent Weight: 200mg |
1495.00 NOK
Pack of 30 |
In Stock
Log in to see stock. |
|||
12728212
|
Macherey-Nagel™
730741 |
3 mL | 50 to 60 μm | Adsorbent Weight: 500mg |
2175.00 NOK
Pack of 30 |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||
12787720
|
Macherey-Nagel™
730735 |
3 mL | 55 to 60 Å | Adsorbent Weight: 60mg |
1335.00 NOK
Pack of 30 |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||
12798202
|
Macherey-Nagel™
730737 |
6 mL | 55 to 60 Å | Adsorbent Weight: 150mg |
2730.00 NOK
Pack of 30 |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||
12738212
|
Macherey-Nagel™
730743 |
6 mL | 50 to 60 μm | Adsorbent Weight: 500mg |
3090.00 NOK
Pack of 30 |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||
Specifications
Spherical PS/DVB Copolymer | |
85 μm | |
Solid Phase Extraction Column |
SPE | |
Chromabond HR-XCW | |
850 m2/g |