Learn More
Chlorotri-n-butylsilane, 97%
Product Code. p-7043920
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
CAS: 995-45-9 | C12H27ClSi | 234.883 g/mol
Chlorotri-n-butylsilane is as silylating agent. It can react with bromo-trifluoro-methane, to product tributyl-trifluoromethyl-silane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chlorotri-n-butylsilane is as silylating agent. It can react with bromo-trifluoro-methane, to product tributyl-trifluoromethyl-silane.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 995-45-9
Molecular Formula C12H27ClSi
Molecular Weight (g/mol) 234.883
MDL Number MFCD00000509
InChI Key JSQJUDVTRRCSRU-UHFFFAOYSA-N
Synonym silane, tributylchloro, chlorotributylsilane, chlorotri-n-butylsilane, tributyl chloro silane, tri-n-butylchlorosilane, tributylsilyl chloride, tributylchloro-silan, chloro-tri-n-butylsilane
PubChem CID 70448
IUPAC Name tributyl(chloro)silane
SMILES CCCC[Si](CCCC)(CCCC)Cl
Specifications
Boiling Point 93°C to 94°C (4mmHg)
Quantity 5 g
UN Number UN2987
Beilstein 1742537
Flash Point 104°C (219°F)
Solubility Information Reacts with water.
Formula Weight 234.89
Percent Purity 97%
Refractive Index 1.446
Sensitivity Moisture sensitive
Density 0.883
Chemical Name or Material Chlorotri-n-butylsilane