Learn More
Thermo Scientific™ Cheminert™ Model C2 Microbore Injectors
Use Thermo Scientific™ Cheminert™ Model C2 Microbore Injectors as injectors or switching valves.
22601.88 NOK
Specifications
Volume (Metric) Sample | 5 μL Loop |
---|---|
For Use With (Equipment) | HPLC |
Type | Microbore Injector/Switching Valve |
Product Code | Brand | Stator Material | Actuation | No. of Ports | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Stator Material | Actuation | No. of Ports | Price | Quantity & Availability | |||
10289274
|
Thermo Scientific™
C2-1000EP |
N60 Stainless | Microelectric | 10 Ports | N/A | ||||
Description
Use Thermo Scientific™ Cheminert™ Model C2 Microbore Injectors as injectors or switching valves. It can be actuated manually or with a microelectronic actuator. With 1/16 in. fittings and 0.010 in. ports, it is available in six-port or ten-port configurations and includes a 5μL sample loop.
- 1/16 in. fittings
- 0.010 in. ports
- Available in 6-port or 10-port configurations
- Available with manual or microelectric actuator
Specifications
5 μL Loop | |
HPLC | |
Microbore Injector/Switching Valve |