Learn More
Bochem™ Chemical Scoop
Manufactured from 18/10 stainless steel. Bochem™ Chemical Scoop has a round barrel.
203.00 NOK - 310.00 NOK
Product Code | Brand | Capacity (Metric) | Length (Metric) | Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Length (Metric) | Size | Price | Quantity & Availability | |||
10653161
|
Bochem™
8803 |
1500 mL | 350 mm | 120 mm |
310.00 NOK
Each |
Estimated Shipment: 01-10-2025
Log in to see stock. |
|||
10569271
|
Bochem™
8800 |
120 mL | 190 mm | 90 mm |
203.00 NOK
Each |
Estimated Shipment: 15-09-2025
Log in to see stock. |
|||
Description
- Round barrel
- High sides
- 18/10 stainless steel
Specifications
Chemical Scoop | |
Stainless Steel (18/10) |