Learn More
Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers
Record seven days of temperature performance at temperatures to -150° C.
Brand: Thermo Scientific™ FCR600TSXLE
23631.54 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Record seven days of temperature performance at temperatures to -150° C using Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers. Recorders use 6 in. (15cm) chart paper and are available as factory or field installed.
Field-installed ink chart recorderServices
Warranty varies by country, please contact us for details
Specifications
Inkless Chart Recorder, Field Installed | |
TSX600 Ultra-low Freezers | |
Recorder |