Learn More
CES1 Mouse anti-Human, Unconjugated, Clone: 1C11C1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67079-1-IG-20UL
 View more versions of this product
Product Code. 16869495
  • 1696.32 NOK / 20µL
Explore more special offers
Description
Description
The CES1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CES1. This antibody recognizes human antigen. The CES1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
CES1
Monoclonal
700 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
1C11C1
Unconjugated
20 μL
1066
IgG1