Learn More
CES1 Mouse anti-Human, Unconjugated, Clone: 1C11C1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67079-1-IG-150UL
This item is not returnable.
View return policy
Description
The CES1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CES1. This antibody recognizes human antigen. The CES1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
CES1 | |
Monoclonal | |
700 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
1C11C1 | |
Unconjugated | |
150 μL | |
1066 | |
IgG1 |