Learn More
Sigma Laborzentrifugen™ Compact Centrifuge
Use this compact centrifuge in any laboratory where space is at a premium. Sigma™ Compact Centrifuges are among the world’s smallest swing-out centrifuges and features many bucket options covering standard tubes. Ideal for small and middle sample sizes.
5180.00€ - 10335.00€
Specifications
Rotor Included | Yes |
---|---|
Type | Centrifuge |
Ventilated | Yes |
Electrical Requirements | 230 V 50 Hz |
Product Code | Brand | Depth (Metric) | Max. Speed | Capacity | Noise Level | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Depth (Metric) | Max. Speed | Capacity | Noise Level | Price | Quantity & Availability | ||
10561315
|
Sigma Laborzentrifugen™
10155 |
452 mm | 15,000 rpm | 4 x 100 mL | <67 dB | ||||
13455145
|
Sigma Laborzentrifugen™
10355 |
600 mm | 15,300 rpm | 4 x 400 mL | 60 dB |
7465.00€
Each |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||
13445155
|
Sigma Laborzentrifugen™
10380 |
650 mm | 13,500 rpm | 4 x 750 mL | 73 dB |
10335.00€
Each |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||
Description
- Stainless steel bowl
- Imbalance switch
- Brushless drive practically maintenance-free
- Double lid locking device with latches
- Automated opening of lid; switchable
- Audible alarm
- No need to open the casing for emergency lid lock release
- Window in the centrifuge lid for external speed calibration
- Produced according to national and international safety regulations (e.g. IEC 1010)
Specifications
Yes | |
Centrifuge | |
Yes | |
230 V 50 Hz |