Learn More
CD52 Mouse anti-Human, Unconjugated, Clone: 1B10E8, Proteintech
Mouse Monoclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | CD52 |
---|---|
Clone | 1B10E8 |
Concentration | 1000 μg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16888975
|
Proteintech
66784-1-IG-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16878975
|
Proteintech
66784-1-IG-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD52 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD52. This antibody recognizes human antigen. The CD52 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
CD52 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
1043 | |
Primary |
1B10E8 | |
Western Blot, ELISA | |
Unconjugated | |
Human | |
IgG1 |