Learn More
Heidolph™ Carousel Attachment
Spare attachment for 12 vessels
Brand: Heidolph™ 549-03000-00
1087.99 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Attachment for 12 vessels with diameters from 16 to 32 mm
Specifications
Test Tubes | |
12 |